Learn More
Med Vet International Vetivex Veterinary pHyLyte Injection pH 7.4 (Multiple Electrolytes), 3000mL
Catalog No. 503045479
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Veterinary pHyLyte Injection pH 7.4 (Multiple Electrolytes Injection, Type 1, USP) is indicated as a source of water and electrolytes or as an alkalinizing agent. See package insert for complete product details.
Catalog No. 50-304-5479 Supplier Med Vet International Supplier No. RXRINGL3000
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Veterinary pHyLyte Injection pH 7.4 (Multiple Electrolytes Injection, Type 1, USP) is indicated as a source of water and electrolytes or as an alkalinizing agent. See package insert for complete product details.