Learn More
phyproof™ 2″-Galloylhyperin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 53209-27-1 |
| Product Line | phyproof |
| Linear Formula | C28H24O16 |
| MDL Number | MFCD07781428 |
| Synonym | 3,3′,4′,5,7-Pentahydroxyflavone 3-beta-galactoside 2′′-gallate; Hyperoside 2′′-gallate; Quercetin 3-beta-galactoside 2′′-gallate |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 616.48 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.