Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ Acetyleugenol, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111001823 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1823 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1823 Supplier MilliporeSigma Supelco Supplier No. PHL82460100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 93-28-7
Product Line phyproof
Density 1.079 g/mL (at 25°C)
Flash Point 110°C - closed cup
Linear Formula C12H14O3
MDL Number MFCD00026191
Boiling Point 281°C to 286°C
Synonym Eugenyl acetate; 4-Allyl-2-methoxyphenyl acetate; O-Acetyleugenol; Eugenol acetate
Type Reference Substance
Quantity 100 mg
Formula Weight 206.24
Percent Purity ≥90% (GC)
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.