Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ Aloe-emodin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002069 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2069 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2069 Supplier MilliporeSigma Supelco Supplier No. PHL8915720MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aloe-emodin, an anthraquinone derivative, is an active constituent of several medicinal plants. It shows gastrointestinal effects by exerting laxative activity.

TRUSTED_SUSTAINABILITY

Specifications

CAS 481-72-1
Product Line phyproof
Linear Formula C15H10O5
MDL Number MFCD00017373
Synonym 1,8-Dihydroxy 3-hydroxymethylanthraquinone; 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone; 3-Hydroxymethylchrysazine; Rhabarberone
Type Reference Substance
Quantity 20 mg
Formula Weight 270.24
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.