Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ Benzyl alcohol, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002445 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2445 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2445 Supplier MilliporeSigma Supelco Supplier No. PHL89164100MG
Add to Cart
Add to Cart

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-51-6
Product Line phyproof
Ignition Point 817°F
Vapor Pressure 3.75 mmHg (77°C)
Vapor Density 3.7 (vs. air)
Density 1.045 g/mL (at 25°C)
Flash Point 101°C - DIN 51758
Linear Formula C6H5 CH2 OH
MDL Number MFCD00004599
Melting Point -16°C to -13°C
Boiling Point 203°C to 205°C
Synonym Benzenemethanol
Type Reference Substance
Quantity 100 mg
Formula Weight 108.14
Percent Purity ≥98% (GC)
Grade Primary Reference Standard
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.