Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ (-)-Caryophyllene oxide, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002398 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2398 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2398 Supplier MilliporeSigma Supelco Supplier No. PHL82496100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 1139-30-6
Product Line phyproof
Flash Point 110°C - closed cup
Linear Formula C15H24 O
MDL Number MFCD00134216
Melting Point 62°C to 63°C
Synonym beta-Caryophyllene epoxide; (-)-Epoxycaryophyllene; (-)-Epoxydihydrocaryophyllene; (1 R,4 R,6 R,10 S)-9-Methylene-4,12,12-trimethyl-5-oxatricyclo[8.2.0.04,6 ]dodecane; trans-Caryophyllene oxide
Type Reference Substance
Quantity 100 mg
Formula Weight 220.35
Percent Purity ≥90% (GC)
Show More Show Less

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.