Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ Gymnemagenin, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002240 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2240 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2240 Supplier MilliporeSigma Supelco Supplier No. PHL8046610MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 22467-07-8
Product Line phyproof
Linear Formula C30H50O6
MDL Number MFCD06656306
Synonym Olean-12-ene-3 beta,16 beta,21 beta,22 alpha,23,28-hexol
Type Reference Substance
Quantity 10 mg
Formula Weight 506.71
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.