Learn More
phyproof™ Malvidin 3-glucoside chloride, ≥95% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 7228-78-6 |
| Product Line | phyproof |
| Linear Formula | C23H25 ClO12 |
| MDL Number | MFCD00185576 |
| Synonym | Oenin chloride; Cyclamin chloride; Enin chloride; Malvidin 3-beta-D-glucopyranoside; Malvidin-3-O-glucoside chloride |
| Type | Reference Substance |
| Quantity | 10 mg |
| Formula Weight | 528.89 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.