Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ Pelargonidin chloride, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002352 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2352 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2352 Supplier MilliporeSigma Supelco Supplier No. PHL8008410MG
Add to Cart
Add to Cart

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 134-04-3
Product Line phyproof
Linear Formula C15H11 ClO5
MDL Number MFCD00017589
Synonym 3,4′,5,7-Tetrahydroxyflavylium chloride; 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-1-benzopyrylium chloride; Pelargonidol chloride
Type Reference Substance
Quantity 10 mg
Formula Weight 306.70
Percent Purity ≥90% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.