Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ Quercitrin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002021 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2021 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2021 Supplier MilliporeSigma Supelco Supplier No. PHL8934620MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 522-12-3
Product Line phyproof
Linear Formula C21H20O11
MDL Number MFCD00016932
Synonym 3,3′,4′,5,7-Pentahydroxyflavone 3-rhamnoside; 3-O-Rhamnosylquercetin; Quercetin 3-rhamnoside; Quercimelin; Quercitroside
Type Reference Substance
Quantity 20 mg
Formula Weight 448.38
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.