Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ trans-m-Coumaric acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002320 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2320 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2320 Supplier MilliporeSigma Supelco Supplier No. PHL83258100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 14755-02-3
Product Line phyproof
Linear Formula HOC6H4 CH=CHCO2 H
MDL Number MFCD00004390
Melting Point 193°C to 195°C
Synonym trans-3-Hydroxycinnamic acid; (E)-3-(3-Hydroxyphenyl)acrylic acid; m-Coumaric acid
Type Reference Substance
Quantity 100 mg
Formula Weight 164.16
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.