Learn More
CPC Scientific Glp-Ala-Asp-Pro-Asn-Lys-Phe-Tyr-Gly-Leu-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP122A
Encompass
Poor inhibitor of water intake that lowers arterial blood pressure, exerts a potent hypotensive action and stimulates extravascular smooth muscle. ONE-LETTER SEQUENCE: Glp-ADPNKFYGLM-NH2MOLECULAR FORMULA: C58H84N14O16S1MOLECULAR WEIGHT:1265.46STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [2507-24-6], RESEARCH AREA: NeuropeptidesREFERENCES: V. Erspamer et al., Experientia, 20, 489 (1964)
Catalog No. 50-210-9956