Learn More
Medchemexpress LLC Physalin B | 23133-56-4 | 1 MG
SDP
Catalog No. 5000417726
Encompass_Preferred
Physalin B, one of the major active steroidal constituents of Cape gooseberry, induces cell cycle arrest and triggers apoptosis in breast cancer cells through modulating p53-dependent apoptotic pathway. It also inhibits the ubiquitin-proteasome pathway and induces incomplete autophagic response in human colon cancer cells in vitro.
  • One of the major active steroidal constituents of Cape gooseberry
  • Induces cell cycle arrest
  • Triggers apoptosis in breast cancer cells
  • Modulates p53-dependent apoptotic pathway
  • Inhibits the ubiquitin-proteasome pathway
  • Induces incomplete autophagic response in human colon cancer cells in vitro
Catalog No. 50-004-17726 Supplier Medchemexpress LLC Supplier No. HYN76951MG
Physalin B, one of the major active steroidal constituents of Cape gooseberry, induces cell cycle arrest and triggers apoptosis in breast cancer cells through modulating p53-dependent apoptotic pathway. It also inhibits the ubiquitin-proteasome pathway and induces incomplete autophagic response in human colon cancer cells in vitro.
  • One of the major active steroidal constituents of Cape gooseberry
  • Induces cell cycle arrest
  • Triggers apoptosis in breast cancer cells
  • Modulates p53-dependent apoptotic pathway
  • Inhibits the ubiquitin-proteasome pathway
  • Induces incomplete autophagic response in human colon cancer cells in vitro