Learn More
TARGETMOL CHEMICALS INC PHYSALIN F 5MG
SDP
Catalog No. 502579048
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Physalin F is a natural blocker of CaV2.3 (R-type) and CaV2.2 (N-type) voltage-gated calcium channels.It is a secosteroid with potent anti-inflammatory and immunomodulatory activities. Physalin F induces apoptosis of PBMC decreasing the spontaneous proliferation and cytokine production caused by Human T-lymphotropic virus type 1 (HTLV-1) infectionpurity: 99%
Catalog No. 50-257-9048 Supplier TARGETMOL CHEMICALS INC Supplier No. T87165MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Physalin F is a natural blocker of CaV2.3 (R-type) and CaV2.2 (N-type) voltage-gated calcium channels.It is a secosteroid with potent anti-inflammatory and immunomodulatory activities. Physalin F induces apoptosis of PBMC decreasing the spontaneous proliferation and cytokine production caused by Human T-lymphotropic virus type 1 (HTLV-1) infectionpurity: 99%