Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Physcion 8-O-β-D-glucopyranoside | 23451-01-6 | 5 MG
SDP

Catalog No. 5000419596
Encompass_Preferred

Physcion 8-O-β-D-glucopyranoside is an anthraquinone compound isolated from Rumex japonicus Houtt. It exhibits anti-inflammatory and anti-cancer properties and can be used for common malignancy cancer research.

  • Anthraquinone compound
  • Isolated from Rumex japonicus Houtt
  • Exhibits anti-inflammatory properties
  • Exhibits anti-cancer properties
  • Can be used for common malignancy cancer research
  • High purity
  • Molecular weight: 446.40
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Melting point: 235°C
  • Relative density: 1.567 g/cm³

Catalog No. 50-004-19596 Supplier Medchemexpress LLC Supplier No. HYN50915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Physcion 8-O-β-D-glucopyranoside is an anthraquinone compound isolated from Rumex japonicus Houtt. It exhibits anti-inflammatory and anti-cancer properties and can be used for common malignancy cancer research.

  • Anthraquinone compound
  • Isolated from Rumex japonicus Houtt
  • Exhibits anti-inflammatory properties
  • Exhibits anti-cancer properties
  • Can be used for common malignancy cancer research
  • High purity
  • Molecular weight: 446.40
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Melting point: 235°C
  • Relative density: 1.567 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.