Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Physcion, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014435 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4435 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4435 Supplier MilliporeSigma Supelco Supplier No. 9389310MG
Add to Cart
Add to Cart
Physcion is an anthraquinone derivative and is available as the active constituent of Chinese medicinal plants such as Rhubarb, Polygonum cuspidatum and Reynoutria multiflora. It exhibits various biological activities such as antitumor, antimicrobial, antifungal, antioxidant and antihuman cytomegalovirus activity.
TRUSTED_SUSTAINABILITY

Specifications

CAS 521-61-9
Physical Form Neat
Linear Formula C16H12O5
MDL Number MFCD00017374
Synonym 1,8-Dihydroxy-3-methoxy-6-methylanthraquinone; 6-O-Methylemodin; 6-Methoxychrysophanic acid; Emodin-3-methyl ether; Parietin; Rheochrysidin
Type Analytical Standard
Quantity 10 mg
Formula Weight 284.26
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.