Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Physical Properties of Glass
GREENER_CHOICESCI_ED

Catalog No. S96641 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96641 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96641 Supplier Innovating Science™ Supplier No. IS9005
Add to Cart
Add to Cart

Includes

Teacher's manual and student guide, analysis copy-masters, one bottle of borosilicate glass beads, one bottle of flint glass beads, one bottle of soda-lime glass beads, one set of refractive index solutions, 15 magnifiers. Enough materials for 15 tests.

Requires

Balance, graduated cylinders (10mL), test tubes

See how forensic scientists use the chemical and physical properties of glass to discover evidence.

Police and crime scene investigators must use all available tools and evidence, working backwards to create the most likely scenario as to what might have occurred. Different types of evidence provide different pieces to the puzzle.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Forensic scientists use the chemical and physical properties of glass to discover evidence
Includes Teacher's manual and student guide, analysis copy-masters, one bottle of borosilicate glass beads, one bottle of flint glass beads, one bottle of soda-lime glass beads, one set of refractive index solutions, 15 magnifiers, enough materials for 15 tests
Quantity 1
Requires Balance, graduated cylinders (10mL), test tubes
Dimensions (L x W x H) 9.75 x 6.75 x 3 in.
Class Size 15 Lab Groups
Product Type Physical Properties of Glass Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.