Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Physics and Chemistry Bulletin Board Charts
SCI_ED

Catalog No. S08984 Shop All NewPath Learning Products
Change view
Click to view available options
Product Type:
Electromagnetism Bulletin Board Chart
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S08984 Electromagnetism Bulletin Board Chart
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08984 Supplier NewPath Learning™ Supplier No. 944610

missing translation for 'validMainframeMsg'

Suited for the reinforcement and review of standards-based topics.

  • Double-sided and laminated
  • Front features graphic overview of topic
  • Reverse provides write-on/wipe-off activities

Specifications

Product Type Electromagnetism Bulletin Board Chart
For Use With (Application) Perfect for the reinforcement and review of standard based topics, provides write-on/wipe-off activities
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade 6 to 12
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.