Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NewPath Learning™ Physics and Chemistry Bulletin Board Charts

Click to view available options
Product Type:
Electromagnetism Bulletin Board Chart
Description
- Double-sided and laminated
- Front features graphic overview of topic
- Reverse provides write-on/wipe-off activities
Specifications
Specifications
Product Type | Electromagnetism Bulletin Board Chart |
Grade | 6 to 12 |
Subject | Science |
For Use With (Application) | Perfect for the reinforcement and review of standard based topics, provides write-on/wipe-off activities |
Certifications/Compliance | Academic Standard for California, Florida, National, New York, Texas |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction