Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Physics with Video Analysis
SDPSCI_ED

Catalog No. S15967ND Shop All Vernier Software Products
Change view
Click to view available options
Format: Printed Lab Book
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S15967ND Format: Printed Lab Book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15967ND Supplier Vernier™ Supplier No. PVA

Includes

Physics with Video Analysis

This book contains a wide selection of video analysis activities for introductory physics at either the high school or college level.

  • 33 ready-to-use student activities, including videos
  • Videos included with Logger Pro
  • Access to up-to-date versions of the activities
  • Instructions for video data analysis with Logger Pro
  • Activities can be used in class, lab, or as independent homework, and do not require any laboratory equipment
  • Essential instructor information including teaching tips, suggested answers, and sample analysis of videos
  • Word-processing files of the student experiments, so you can edit the files to match your teaching preferences
  • A generous site license—buy one book and duplicate the experiments for your class
  • Printed copy of book
  • Covers topics including kinematics, dynamics, circuits, sound, electrostatics, and more.

Warranty and Services

5-year limited warranty

Specifications

Grade 9 to 12
Description ISBN 978-1-929075-11-9
Format Printed Lab Book
Languages English
For Use With (Application) Educational use only
Subject Physics
Author Priscilla Laws, Robert Teese, Maxine Willis, and Patrick Cooney
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.