Learn More
AdipoGen Physiological Saline [Sodium Chloride 0.9%] Endotoxin-free Sterile Solution, 100 ml
SDP
Catalog No. 501490621
Encompass_Preferred
Physiological Saline [Sodium Chloride 0.9%] Endotoxin-free Sterile Solution (100 ml), AdipoGen Life Sciences. Chemical. Use for the dilution of TLRpureTM LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).
Catalog No. 50-149-0621 Supplier AdipoGen Supplier No. IAX900003L100
Physiological Saline [Sodium Chloride 0.9%] Endotoxin-free Sterile Solution (100 ml), AdipoGen Life Sciences. Chemical. Use for the dilution of TLRpureTM LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).