Learn More
AdipoGen Physiological Saline (Sodium Chloride 0.9%) Endotoxin-free Sterile Solution, 10 mL
SDP
Catalog No. 50596732
Encompass_Preferred
Synonyms: Sodium Chloride 0.9%. Use for the dilution of TLRpureTM LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).
Catalog No. 50-596-732 Supplier AdipoGen Supplier No. IAX900003L010
Synonyms: Sodium Chloride 0.9%. Use for the dilution of TLRpureTM LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).