Learn More
AdipoGen Physiological Saline (Sodium Chloride 0.9%) Endotoxin-free Sterile Solution, 3x10 mL
SDP
Catalog No. 501017111
Encompass_Preferred
Synonyms: Sodium Chloride 0.9%. Use for the dilution of TLRpureTM LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).
Catalog No. 50-101-7111 Supplier AdipoGen Supplier No. IAX9000033010
Synonyms: Sodium Chloride 0.9%. Use for the dilution of TLRpureTM LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).