Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Physostigmine free base, TCI America™
SDP

Catalog No. P0406200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0406200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0406200MG Supplier TCI America Supplier No. P0406200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 57-47-6
Molecular Formula C15H21N3O2
Molecular Weight (g/mol) 275.352
MDL Number MFCD00151090
InChI Key PIJVFDBKTWXHHD-HIFRSBDPSA-N
Synonym Eserine
PubChem CID 5983
ChEBI CHEBI:27953
IUPAC Name [(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-methylcarbamate
SMILES CC12CCN(C1N(C3=C2C=C(C=C3)OC(=O)NC)C)C

Specifications

Physical Form Powder/Crystals
UN Number 1544
Color White/Yellow
Melting Point 105°C
Quantity 200 mg
Formula Weight 275.35
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Physostigmine free base
RTECSNumber TJ2100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.