Learn More
Medchemexpress LLC Physostigmine salicylate | 57-64-7 | 99.7% | 50 MG
SDP
Catalog No. 5000412242
Encompass_Preferred
Physostigmine salicylate (Eserine salicylate) is a reversible acetylcholinesterase (AChE) inhibitor. It can cross the blood-brain barrier and stimulate central cholinergic neurotransmission. Physostigmine salicylate has been shown to reverse memory deficits in transgenic mice with Alzheimer's disease and also acts as an antidote for anticholinergic poisoning.
  • Reversible acetylcholinesterase (AChE) inhibitor
  • Crosses the blood-brain barrier
  • Stimulates central cholinergic neurotransmission
  • Can reverse memory deficits in transgenic mice with Alzheimer's disease
  • Antidote for anticholinergic poisoning
Catalog No. 50-004-12242 Supplier Medchemexpress LLC Supplier No. HYB126650MG
Add to Cart
Add to Cart
Physostigmine salicylate (Eserine salicylate) is a reversible acetylcholinesterase (AChE) inhibitor. It can cross the blood-brain barrier and stimulate central cholinergic neurotransmission. Physostigmine salicylate has been shown to reverse memory deficits in transgenic mice with Alzheimer's disease and also acts as an antidote for anticholinergic poisoning.
  • Reversible acetylcholinesterase (AChE) inhibitor
  • Crosses the blood-brain barrier
  • Stimulates central cholinergic neurotransmission
  • Can reverse memory deficits in transgenic mice with Alzheimer's disease
  • Antidote for anticholinergic poisoning