Learn More
TARGETMOL CHEMICALS INC Physostigmine Salicylate 5MG
SDP
Catalog No. 502880709
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Physostigmine, a parasympathomimetic alkaloid, is a reversible cholinesterase inhibitor. It extracts from the Calabar bean. Purity 99.36%
Catalog No. 50-288-0709 Supplier TARGETMOL CHEMICALS INC Supplier No. T01305MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Physostigmine, a parasympathomimetic alkaloid, is a reversible cholinesterase inhibitor. It extracts from the Calabar bean. Purity 99.36%