Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phytane, ≥95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015018 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5018 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5018 Supplier MilliporeSigma Supelco Supplier No. 8016525MG
Add to Cart
Add to Cart

Specifications

CAS 638-36-8
Density 0.791 g/mL (at 20°C)
Physical Form Neat
Linear Formula C20H42
MDL Number MFCD00060946
Boiling Point 69°C to 71°C (0.001 mmHg)
Synonym 2,6,10,14-Tetramethylhexadecane
Type Analytical Standard
Quantity 25 mg
Formula Weight 282.55
Percent Purity ≥95% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.