Learn More
Medchemexpress LLC Phytanic acid-d3 | 1383920-40-8 | 99.1% | 315.55 | 1 MG
SDP
Catalog No. 5000437728
Encompass_Preferred
Phytanic acid-d3 is the deuterium-labeled Phytanic acid. It is an endogenous metabolite found in blood. This compound can be utilized as a tracer and an internal standard for quantitative analysis, and is also explored for its potential impact on pharmacokinetic and metabolic profiles.
  • Deuterium-labeled form of Phytanic acid
  • Useful for research related to Zellweger Syndrome, Alpha Methylacyl CoA Racemase Deficiency, Rhizomelic Chondrodysplasia Punctata, and Infantile Refsum Disease
  • Can serve as a tracer in experiments
  • Acts as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Potential to influence pharmacokinetic and metabolic profiles of drugs through deuterium substitution
Catalog No. 50-004-37728 Supplier Medchemexpress LLC Supplier No. HY113067S1MG
Phytanic acid-d3 is the deuterium-labeled Phytanic acid. It is an endogenous metabolite found in blood. This compound can be utilized as a tracer and an internal standard for quantitative analysis, and is also explored for its potential impact on pharmacokinetic and metabolic profiles.
  • Deuterium-labeled form of Phytanic acid
  • Useful for research related to Zellweger Syndrome, Alpha Methylacyl CoA Racemase Deficiency, Rhizomelic Chondrodysplasia Punctata, and Infantile Refsum Disease
  • Can serve as a tracer in experiments
  • Acts as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Potential to influence pharmacokinetic and metabolic profiles of drugs through deuterium substitution