Learn More
Medchemexpress LLC Phytanic acid-d3 | 1383920-40-8 | 99.1% | 315.55 | C20H37D3O2 | 5 MG
SDP
Catalog No. 5000264335
Encompass_Preferred
Phytanic acid-d3 is a deuterium-labeled analogue of phytanic acid used as an internal standard for accurate quantitation by GC- or LC-MS in biochemical and clinical research. It has molecular formula C20H37D3O2, molecular weight 315.55, and CAS 1383920-40-8, and is supplied in research-scale quantities for analytical use.
  • Intended for use as an internal standard in GC- or LC-MS assays.
  • Deuterium labeling provides a distinct mass shift for reliable differentiation from the native analyte.
  • Coelutes with native phytanic acid to support accurate quantitation.
  • Offered in small research-scale quantities suitable for method development and standard curves.
  • High isotopic and chemical purity supports precise measurement.
Catalog No. 50-002-64335 Supplier Medchemexpress LLC Supplier No. HY113067S5MG
Phytanic acid-d3 is a deuterium-labeled analogue of phytanic acid used as an internal standard for accurate quantitation by GC- or LC-MS in biochemical and clinical research. It has molecular formula C20H37D3O2, molecular weight 315.55, and CAS 1383920-40-8, and is supplied in research-scale quantities for analytical use.
  • Intended for use as an internal standard in GC- or LC-MS assays.
  • Deuterium labeling provides a distinct mass shift for reliable differentiation from the native analyte.
  • Coelutes with native phytanic acid to support accurate quantitation.
  • Offered in small research-scale quantities suitable for method development and standard curves.
  • High isotopic and chemical purity supports precise measurement.