Learn More
Selleck Chemical LLC Phytic acid-25mg
SDP
Supplier:  Selleck Chemical LLC S379325MG
Encompass_Preferred
Phytic acid (Inositol polyphosphate, Inositol hexakisphosphate) is the principal storage form of phosphorus in many plant tissues, especially bran and seeds. It can act as a cofactor in DNA repair by nonhomologous end-joining. Phytic acid inhibits the enzymatic superoxide source xanthine oxidase (XO), and has antioxidative, neuroprotective, anti-inflammatory effects.
Catalog No. 50-238-9850