Learn More
Apexbio Technology LLC Phytic acid 83-86-3 250mg
SDP
Catalog No. 5000090336
Encompass_Preferred
Phytic acid (CAS 83-86-3) is a naturally occurring polyphosphorylated carbohydrate predominantly isolated from plant seeds It functions as a chelating agent binding mineral ions such as iron and calcium thereby modulating their bioavailability Through its antioxidant properties and regulation of cellular signaling pathways phytic acid is investigated for roles in cell proliferation apoptosis and metabolism Its utility extends to studies on mineral absorption cancer biology and oxidative stress The compound is soluble in water ethanol and acetone and it should be stored in a tightly sealed container under cool dry conditions to maintain stability
Catalog No. 50-000-90336 Supplier Apexbio Technology LLC Supplier No. N1645250
Phytic acid (CAS 83-86-3) is a naturally occurring polyphosphorylated carbohydrate predominantly isolated from plant seeds It functions as a chelating agent binding mineral ions such as iron and calcium thereby modulating their bioavailability Through its antioxidant properties and regulation of cellular signaling pathways phytic acid is investigated for roles in cell proliferation apoptosis and metabolism Its utility extends to studies on mineral absorption cancer biology and oxidative stress The compound is soluble in water ethanol and acetone and it should be stored in a tightly sealed container under cool dry conditions to maintain stability