Learn More
Selleck Chemical LLC Phytic acid dipotassium salt S3326-100mg
SDP
Catalog No. 503108744
Encompass_Preferred
Phytic acid dipotassium salt (Phytic acid potassium IP6 (PS)) an endogenous metabolite in rice grain extract and digest inhibits -secretase 1 (BACE1) with IC50 of approximately 0 25 M
Catalog No. 50-310-8744 Supplier Selleck Chemical LLC Supplier No. S3326100MG
Phytic acid dipotassium salt (Phytic acid potassium IP6 (PS)) an endogenous metabolite in rice grain extract and digest inhibits -secretase 1 (BACE1) with IC50 of approximately 0 25 M