Learn More
Medchemexpress LLC HY-N0814A 250mg Medchemexpress, Phytic acid (dodecasodium salt hydrate) CAS:123408-98-0 Purity:>98%
SDP
Catalog No. 501933730
Encompass_Preferred
Medchemexpress, HY-N0814A 250mg Phytic acid (dodecasodium salt hydrate) CAS:123408-98-0 Phytic acid dodecasodium salt hydrate is a phosphorus storage compound of seeds and cereal grains. Phytic acid dodecasodium salt hydrate is known as a food inhibitor, which has a strong ability to chelate multivalent metal ions, specially zinc, calcium, iron and as with protein residue. Phytic acid dodecasodium salt hydrate inhibits the enzymatic superoxide source xanthine oxidase (XO) , and has antioxidative, neuroprotective, anti-inflammatory effects [1] [2] [3] [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3730 Supplier Medchemexpress LLC Supplier No. HYN0814A250MG
Medchemexpress, HY-N0814A 250mg Phytic acid (dodecasodium salt hydrate) CAS:123408-98-0 Phytic acid dodecasodium salt hydrate is a phosphorus storage compound of seeds and cereal grains. Phytic acid dodecasodium salt hydrate is known as a food inhibitor, which has a strong ability to chelate multivalent metal ions, specially zinc, calcium, iron and as with protein residue. Phytic acid dodecasodium salt hydrate inhibits the enzymatic superoxide source xanthine oxidase (XO) , and has antioxidative, neuroprotective, anti-inflammatory effects [1] [2] [3] [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.