Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phytic acid potassium | 129832-03-7 | MFCD00133773 | 98.0% | 736.22 g/mol | C6H16K2O24P6 | 1 G
SDP

Catalog No. 5000299681
Encompass_Preferred

Phytic acid potassium is the dipotassium salt of phytic acid (inositol hexaphosphate), an endogenous inositol phosphate used in biochemical and cellular research as a chelator and metabolic probe. The product is supplied with reported purity, solubility, stability data, and accompanying certificate of analysis and datasheet for quality verification.

  • High purity (98.0%) suitable for research applications.
  • Available in multiple sizes including 1 g for small-scale studies.
  • Soluble in DMSO and water; in vitro solubility reported at ~100 mg/mL.
  • Formulation guidance and in vivo protocols provided for dosing.
  • Store sealed and protected from moisture; solvent storage stability data provided.

Catalog No. 50-002-99681 Supplier Medchemexpress LLC Supplier No. HYW0180591G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Phytic acid potassium is the dipotassium salt of phytic acid (inositol hexaphosphate), an endogenous inositol phosphate used in biochemical and cellular research as a chelator and metabolic probe. The product is supplied with reported purity, solubility, stability data, and accompanying certificate of analysis and datasheet for quality verification.

  • High purity (98.0%) suitable for research applications.
  • Available in multiple sizes including 1 g for small-scale studies.
  • Soluble in DMSO and water; in vitro solubility reported at ~100 mg/mL.
  • Formulation guidance and in vivo protocols provided for dosing.
  • Store sealed and protected from moisture; solvent storage stability data provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.