Learn More
Medchemexpress LLC Phytic acid potassium | 129832-03-7 | MFCD00133773 | 98.0% | 736.22 g/mol | C6H16K2O24P6 | 1 ML
SDP
Catalog No. 5000302043
Encompass_Preferred
Phytic acid potassium is the potassium salt of phytic acid (inositol hexakisphosphate), described as an endogenous metabolite. This item is provided as a ready-to-use 10 mM solution in DMSO (1 mL) with a listed purity of 98.0%, intended for biochemical and metabolic research and analytical applications.
  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL aliquot.
  • High purity (98.0%) for research use.
  • Suitable for biochemical and metabolic studies.
  • Acts as a chelator of divalent and trivalent metal ions.
Catalog No. 50-003-02043 Supplier Medchemexpress LLC Supplier No. HYW01805910MM/1ML
Add to Cart
Add to Cart
Phytic acid potassium is the potassium salt of phytic acid (inositol hexakisphosphate), described as an endogenous metabolite. This item is provided as a ready-to-use 10 mM solution in DMSO (1 mL) with a listed purity of 98.0%, intended for biochemical and metabolic research and analytical applications.
  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL aliquot.
  • High purity (98.0%) for research use.
  • Suitable for biochemical and metabolic studies.
  • Acts as a chelator of divalent and trivalent metal ions.