Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phytic acid solution

Catalog No. 593648250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-364-8250ML 250 mL
59-364-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59-364-8250ML Supplier Sigma Aldrich Supplier No. 593648250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phytic acid is a mineral chelator that can bind to minerals to form mineral-phytate complex.It can also complex with metal ions bound to the surface of magnesium alloy to form a conversion coating, which can improve the resistance of magnesium alloy towards corrosion.

Specifications

CAS 83-86-3
Density 1.432 g/mL (at 25°C)
Boiling Point 105°C
Molecular Formula C6H18O24P6
Linear Formula C6H18O24P6
MDL Number MFCD00082309
Quantity 250 mL
Synonym myo-Inositol hexakis(dihydrogen phosphate)
Molecular Weight (g/mol) 660.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.