Learn More
AdipoGen Phytic acid . sodium salt . hydrate (100 mg)
SDP
Supplier:  AdipoGen AGCN20528M100
Encompass_Preferred
Chemical. CAS: 14306-25-3 (anhydrous). Formula: C6H12O24P6 . xNa . yH2O. MW: 660.0 (anhydrous free acid). Isolated from Oryza sativa. Direct activator of necroptosis inducer MLKL (mixed lineage kinase domain-like). IP6 is a phosphate ester of inositol (Ins) and precursor for second messengers in cell signaling. Iron (Fe) Chelator that binds also to Ca, Mg, Zn. Considered as an anti-nutritional component in cereals, seeds and beans that binds these minerals and prevents their absorption in the gastrointestinal tract. Formulations containing phytic acid have been used as preservatives in food production. Antioxidant with anticancer, anti-inflammatory, antidiabetic, neuroprotective, hypocholesterolemic and hypolipidemic effects. Increases insulin sensitivity in adipocytes by increasing lipid storage capacity, improving glucose uptake and inhibiting lipolysis.
Catalog No. 50-205-3703