Learn More
Biotage PhyTip PTH-91-40-03, IMAC Column, 1 mL, 40 μL Nickel-IMAC resin, automated purification of His-tagged proteins, pack size 96 columns, ready-to-use, purification in 15 min
SDP
Catalog No. NC2316438
Encompass
PhyTip PTH-91-40-03, IMAC Column, 1 mL, 40 μL Nickel-IMAC resin, automated purification of His-tagged proteins, pack size 96 columns, ready-to-use, purification in 15 min
Catalog No. NC2316438 Supplier Biotage Supplier No. PTH914003
PhyTip PTH-91-40-03, IMAC Column, 1 mL, 40 μL Nickel-IMAC resin, automated purification of His-tagged proteins, pack size 96 columns, ready-to-use, purification in 15 min