Learn More
Biotage IMAC PhyTip columns, 300 μL volume, 20 μL resin bed, Nickel-IMAC resin, 96-well plate compatible, 96 per pack
SDP
Catalog No. NC3433720
Encompass
IMAC PhyTip columns, 300 μL volume, 20 μL resin bed, Nickel-IMAC resin, 96-well plate compatible, 96 per pack
Catalog No. NC3433720 Supplier Biotage Supplier No. PTX932003
IMAC PhyTip columns, 300 μL volume, 20 μL resin bed, Nickel-IMAC resin, 96-well plate compatible, 96 per pack