Learn More
Biotage PhyTip 500 mL MabSelect PrismA column, 500 μL volume, Protein A PhyTip column, for IgG antibodies, high recovery 70-90%, disposable pipette tip format
SDP
Catalog No. NC2953441
Encompass
The Biotage PhyTip 500 mL MabSelect PrismA columns are advanced pipette tips designed for the automated purification of IgG antibodies. These optimized columns feature dual flow chromatography technology that enables efficient sample concentration and purification, delivering high-purity IgGs rapidly. The use of MabSelect PrismA resin technology ensures enhanced binding capacity and alkaline stability, allowing for robust purification workflows. Integrated protocols facilitate use with automated systems, making processing quicker and more efficient.
  • High purity and recovery achieved from diverse sample matrices
  • Automated compatibility for high-throughput processing of up to 96 samples
  • Integrated sample concentration and purification in a single step
  • Optimized resin technology for enhanced dynamic binding capacity
  • Supplied with pre-validated protocols for seamless integration
  • Format designed for disposable use
Catalog No. NC2953441 Supplier Biotage Supplier No. PTT9540OG
The Biotage PhyTip 500 mL MabSelect PrismA columns are advanced pipette tips designed for the automated purification of IgG antibodies. These optimized columns feature dual flow chromatography technology that enables efficient sample concentration and purification, delivering high-purity IgGs rapidly. The use of MabSelect PrismA resin technology ensures enhanced binding capacity and alkaline stability, allowing for robust purification workflows. Integrated protocols facilitate use with automated systems, making processing quicker and more efficient.
  • High purity and recovery achieved from diverse sample matrices
  • Automated compatibility for high-throughput processing of up to 96 samples
  • Integrated sample concentration and purification in a single step
  • Optimized resin technology for enhanced dynamic binding capacity
  • Supplied with pre-validated protocols for seamless integration
  • Format designed for disposable use