Learn More
Biotage PhyTip Column, 500 μL volume, 40 μL resin, IgG antibody purification, dual flow chromatography, pack of 96, high binding affinity, high recovery rate 70-90%, elution in small volumes
SDP
Catalog No. NC2313936
Encompass
The Biotage Protein A PhyTip columns offer advanced purification solutions for the automated isolation of IgG antibodies from complex biological samples. These high-performance columns utilize patented dual flow chromatography to enhance binding interactions with a packed Protein A resin bed, resulting in improved recovery and purity of purified antibodies. Ideal for high-throughput laboratories, these columns allow for quick processing times while integrating seamlessly with various automated liquid handling systems.
  • Efficient purification of IgG antibodies with high purity from diverse background proteins.
  • Achieves 70-90% recovery of IgGs through a unique packed resin bed design.
  • Purifies up to 96 samples in as little as 15 minutes.
  • Compatible with major automated liquid handling systems, ensuring streamlined workflows.
  • Utilizes high binding affinity and capacity for effective capture of target IgGs.
Catalog No. NC2313936 Supplier Biotage Supplier No. PTT954001
The Biotage Protein A PhyTip columns offer advanced purification solutions for the automated isolation of IgG antibodies from complex biological samples. These high-performance columns utilize patented dual flow chromatography to enhance binding interactions with a packed Protein A resin bed, resulting in improved recovery and purity of purified antibodies. Ideal for high-throughput laboratories, these columns allow for quick processing times while integrating seamlessly with various automated liquid handling systems.
  • Efficient purification of IgG antibodies with high purity from diverse background proteins.
  • Achieves 70-90% recovery of IgGs through a unique packed resin bed design.
  • Purifies up to 96 samples in as little as 15 minutes.
  • Compatible with major automated liquid handling systems, ensuring streamlined workflows.
  • Utilizes high binding affinity and capacity for effective capture of target IgGs.