Learn More
Biotage PHYTIP COLUMNS(0.5ML-VOL)160UL
SDP
Catalog No. NC1770360
Encompass
Box of 96 PhyTip Columns (500ul Volume) containing 160ul of ProPlus affinity resin per column, Tecan MCA
Catalog No. NC1770360 Supplier Biotage Supplier No. PTT951607
Box of 96 PhyTip Columns (500ul Volume) containing 160ul of ProPlus affinity resin per column, Tecan MCA