Learn More
Biotage PHYTIP COLUMNS (200 UL VOLUME)
SDP
Catalog No. NC1485506
Encompass
Box of 96 PhyTip columns (200 μL volume) containing 5 μL of Streptavidin per column for MEA or PureSpeed system.
Catalog No. NC1485506 Supplier Biotage Supplier No. PTR920505
Box of 96 PhyTip columns (200 μL volume) containing 5 μL of Streptavidin per column for MEA or PureSpeed system.