Learn More
Biotage PhyTip Column, Box of 96, for the Agilent Bravo System
SDP
Catalog No. NC1529644
Encompass
Box of 96 PhyTip columns (200 L volume) containing 20 L of ProPlus affinity resin per column for the Agilent Bravo system.
Catalog No. NC1529644 Supplier Biotage Supplier No. PTV922007
Box of 96 PhyTip columns (200 L volume) containing 20 L of ProPlus affinity resin per column for the Agilent Bravo system.