Learn More
Biotage PHYTIP COLUMNS W40UL IMAC
SDP
Catalog No. NC1798097
Encompass
Box of 96 PhyTip columns (1 ml volume) containing 40 µl of IMAC affinity resin per column
Catalog No. NC1798097 Supplier Biotage Supplier No. PTR914003
Box of 96 PhyTip columns (1 ml volume) containing 40 µl of IMAC affinity resin per column