Learn More
Cayman Chemical Phytanic Acid, Purity: ≥96%, CAS: 14721-66-5, 50 mg
SDP
Catalog No. 502895013
Encompass_Preferred
Phytanic acid is a saturated 20-carbon branched-chain fatty acid which can only be derived from dietary sources. In cases of peroxisomal disorders, elevated levels of phytanic acid have been observed, and this is linked to diseases such as infantile phytanic acid storage disease and Refsum’s disease.{1912}
Catalog No. 50-289-5013 Supplier Cayman Chemical Supplier No. 9036050 MG
Phytanic acid is a saturated 20-carbon branched-chain fatty acid which can only be derived from dietary sources. In cases of peroxisomal disorders, elevated levels of phytanic acid have been observed, and this is linked to diseases such as infantile phytanic acid storage disease and Refsum’s disease.{1912}