Learn More
Cayman Chemical PhytnIc AcId-d3 10mg
SDP
Supplier:  Cayman Chemical 2906810 MG
Encompass_Preferred
An internal standard for the quantification of phytanic acid by GC- or LC-MS
Catalog No. 50-294-0571