Learn More
Cayman Chemical Phytanic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1383920-40-8, 1 mg
SDP
Catalog No. 502941345
Encompass_Preferred
An internal standard for the quantification of phytanic acid by GC- or LC-MS
Catalog No. 50-294-1345 Supplier Cayman Chemical Supplier No. 290681 MG
An internal standard for the quantification of phytanic acid by GC- or LC-MS