Learn More
Cayman Chemical Phytanic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1383920-40-8, 5 mg
SDP
Catalog No. 502940087
Encompass_Preferred
An internal standard for the quantification of phytanic acid by GC- or LC-MS
Catalog No. 50-294-0087 Supplier Cayman Chemical Supplier No. 290685 MG
An internal standard for the quantification of phytanic acid by GC- or LC-MS