Learn More
Cayman Chemical Phytantriol, Purity: ≥95%, CAS: 74563-64-7, 100 mg
SDP
Catalog No. 502978659
Encompass_Preferred
An aliphatic alcohol; has been used in the formation of LCNPs and cubosomes; cubosomes containing phytantriol are bactericidal against LPS-deficient and polymyxin-resistant A. baumannii strains; LCNPs containing phytantriol and encapsulating berberine reduce TNF-α and iNOS levels and the production of ROS in LPS-activated RAW 264.7 macrophages
Catalog No. 50-297-8659 Supplier Cayman Chemical Supplier No. 40742100 MG
An aliphatic alcohol; has been used in the formation of LCNPs and cubosomes; cubosomes containing phytantriol are bactericidal against LPS-deficient and polymyxin-resistant A. baumannii strains; LCNPs containing phytantriol and encapsulating berberine reduce TNF-α and iNOS levels and the production of ROS in LPS-activated RAW 264.7 macrophages